missing translation for 'onlineSavingsMsg'
Learn More

Cerium silicide, 99.9% (REO)

Catalog No. p-7050199
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
AA3583606 5 g
AA3583614 25 g
2 options

Catalog No. AA3583606

Supplier: Thermo Scientific Chemicals 03583606

Only null left

CAS: 12014-85-6 | CeSi2 | 196.286 g/mol

Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12014-85-6
Molecular Formula CeSi2
Molecular Weight (g/mol) 196.29
MDL Number MFCD00168082
InChI Key KLTZYOJHFGPJMO-UHFFFAOYSA-N
Synonym cerium silicide, cerium disilicide
PubChem CID 6336874
IUPAC Name λ⁴-cerium(4+) disil-1-ene-1,1,2,2-tetraide
SMILES [Ce+4].[Si--]=[Si--]

Specifications

Melting Point 1620°C
Color Gray
Physical Form Pieces
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 196.29
Percent Purity 99.9%
Odor Odorless
Density 5.31 g/mL
Assay (REO)
Chemical Name or Material Cerium silicide
Show More Show Less
EINECSNumber 234-602-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only