missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium silicide, 99.9% (REO)
Description
Cerium silicide is used as a surrogate material to investigate the feasibility of high-energy ball milling and Spark Plasma Sintering (SPS) for the production of uranium silicide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1620°C |
| Color | Gray |
| Physical Form | Pieces |
| Quantity | 25 g |
| Odor | Odorless |
| Density | 5.31 g/mL |
| Solubility Information | Insoluble in water. |
| Formula Weight | 196.29 |
| Percent Purity | 99.9% |
| Assay | (REO) |
| Show More |
Safety and Handling
| EINECSNumber | 234-602-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only