missing translation for 'onlineSavingsMsg'
Learn More

Cerium powder, -60 mesh, 99.9% (REO)

Catalog No. p-7080008
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4590906 5 g
AA4590914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4590906 Supplier Thermo Scientific Chemicals Supplier No. 04590906
Only null left

CAS: 7440-45-1 | Ce | 140.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Color Silver
Quantity 5 g
Mesh Size −200 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
Form Powder
UN Number UN3078
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Cerium powder
Hazard Category H228-H261
Hazard Statement GHS H Statement
H228-H261
Flammable solid.
In contact with water releases flammable gas.
Precautionary Statement P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: CERIUM
EINECSNumber 231-154-9
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only