missing translation for 'onlineSavingsMsg'
Learn More

Cerium oxide, 20% in H{2}O, colloidal dispersion

Catalog No. AA4012522
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4012522 100 g
AA4012536 500 g
AA40125A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4012522 Supplier Thermo Scientific Chemicals Supplier No. 04012522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Widely applied as a solid electrolyte in fuel cells. Also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. | CAS: 1306-38-3 | CeO2 | 172.114 g/mol

Cerium(IV) oxide is widely applied as a solid electrolyte in fuel cells. It is also used in ceramics and to polish glass and stones. It can also be employed as a co-catalyst in many reactions. It is also used as an antioxidant and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles and sunscreens in combination with other materials. It finds use as a catalyst support and to sensitize photosensitive glass. It is utilized to increase specific surface area and inhibit sintering as well as pore size growth at a high temperature by addition with CuO/TiO2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specifications

Physical Form Particles in Liquid
Quantity 100 g
Solubility Information Immiscible with water.
Formula Weight 172.12
Concentration or Composition (by Analyte or Components) 20% in H2O
Packaging Plastic bottle
Specific Gravity 1.22
Density 1.22
Chemical Name or Material Cerium oxide
EINECSNumber 215-150-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only