missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium molybdenum oxide, 99%
Description
Cerium molybdenum oxide is used in the aerospace and coating industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 973°C |
| Color | Brown |
| Physical Form | Powder |
| Quantity | 5 g |
| Assay Percent Range | 99% |
| Solubility Information | Insoluble in water. |
| Formula Weight | 760.05 |
| Chemical Name or Material | Cerium molybdenum oxide |
Safety and Handling
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only