missing translation for 'onlineSavingsMsg'
Learn More

Cerium molybdenum oxide, 99%

Catalog No. AA4021006
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4021006 5 g
AA4021014 25 g
AA4021022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4021006 Supplier Thermo Scientific Chemicals Supplier No. 04021006

CAS: 13454-70-1 | Ce2Mo3O12 | 760.07 g/mol

Cerium molybdenum oxide is used in the aerospace and coating industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13454-70-1
Molecular Formula Ce2Mo3O12
Molecular Weight (g/mol) 760.07
MDL Number MFCD00798520
InChI Key BPOYBXJKAISYKA-UHFFFAOYSA-N
Synonym Cerium molybdate

Specifications

Melting Point 973°C
Color Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 760.05
Chemical Name or Material Cerium molybdenum oxide
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only