missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) trifluoromethanesulfonate, 98%

Catalog No. AAL2005606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2005606 5 g
AAL2005614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2005606 Supplier Thermo Scientific Chemicals Supplier No. L2005606

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 107792-63-2 | C4CeF12O12S4 | 736.37 g/mol

Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) trifluoromethanesulfonate, is used as a strong oxidizing agent, in the oxidation of benzyl alcohols and alkylbenzenes to aryl aldehydes and aryl ketones respectively. It can also be used as a catalyst for high-yield deprotection of tert-butyldimethylsilyl ethers

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.

Chemical Identifiers

CAS 107792-63-2
Molecular Formula C4CeF12O12S4
Molecular Weight (g/mol) 736.37
MDL Number MFCD03844746
InChI Key BYAJTISDHIASIP-UHFFFAOYSA-J
Synonym cerium iv trifluoromethanesulfonate, cerium iv triflate, cerium trifluoromethanesulfonate, acmc-20alhh, ceric triflate, cerium iv trifluoromethanesulphonate, cerium iv trifluoromethanesulfonate, ?ce cf3so3 4, trifluoromethanesulfonic acid, cerium iv salt, methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1
PubChem CID 9875257
IUPAC Name cerium(4+);trifluoromethanesulfonate
SMILES [Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 736.39
Percent Purity 98%
Chemical Name or Material Cerium(IV) trifluoromethanesulfonate
Hazard Category H314-H500
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only