missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) sulfate, 95%

CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol

Supplier:  Thermo Scientific Chemicals 404341000

 View more versions of this product

Catalog No. AC404341000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium(IV) sulfate
13590-82-4
39.6 to 44.7% (Ce) (Iodometry)
Ce(SO4)2
15,1989
Solubility in water: soluble. Other solubilities: soluble in dilute acids
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
332.23
332.23
195.0°C
100 g
CeO8S2
MFCD00148852
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
VZDYWEUILIUIDF-UHFFFAOYSA-J
cerium(4+);disulfate
159684
95%

Chemical Identifiers

13590-82-4
332.23
VZDYWEUILIUIDF-UHFFFAOYSA-J
159684
[Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
CeO8S2
MFCD00148852
ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate
cerium(4+);disulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 237-029-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only