Learn More
Cerium(IV) sulfate, 95%
CAS: 13590-82-4 | CeO8S2 | 332.23 g/mol
Supplier: Thermo Scientific Chemicals 404340250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Cerium(IV) sulfate | |
| 13590-82-4 | |
| 39.6 to 44.7% (Ce) (Iodometry) | |
| Ce(SO4)2 | |
| 15,1989 | |
| Solubility in water: soluble. Other solubilities: soluble in dilute acids | |
| [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
| 332.23 | |
| 332.23 |
| 195.0°C | |
| 25 g | |
| CeO8S2 | |
| MFCD00148852 | |
| ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
| VZDYWEUILIUIDF-UHFFFAOYSA-J | |
| cerium(4+);disulfate | |
| 159684 | |
| 95% |
Chemical Identifiers
| 13590-82-4 | |
| 332.23 | |
| VZDYWEUILIUIDF-UHFFFAOYSA-J | |
| 159684 | |
| [Ce+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
| CeO8S2 | |
| MFCD00148852 | |
| ceric sulfate, cerium iv sulfate, ceric disulfate, ceric sulphate, cerium disulfate, cerium 4+ sulfate, cerium 4+ disulphate, unii-4f66fi5t7x, sulfuric acid, cerium 4+ salt 2:1, cerium 4+ disulfate | |
| cerium(4+);disulfate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 237-029-5
RUO – Research Use Only