missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, hydrated

Catalog No. p-4824661
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
50 g
Catalog No. Quantity
AA2112018 50 g
AA2112030 250 g
2 options

Catalog No. AA2112018

Supplier: Thermo Scientific Chemicals 02112018

Only null left

CAS: 12014-56-1 | CeH8O4 | 212.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12014-56-1
Molecular Formula CeH8O4
Molecular Weight (g/mol) 212.18
MDL Number MFCD00010927
InChI Key DPUCLPLBKVSJIB-UHFFFAOYSA-N
SMILES O.O.O.O.[Ce]

Specifications

Color Brown to Yellow
Physical Form Lump/Powder
Quantity 50 g
Formula Weight 172.12 (anhydrous)
Odor Odorless
Chemical Name or Material Cerium(IV) oxide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 234-599-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only