missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, hydrated

Catalog No. AA2112018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2112018 50 g
AA2112030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2112018 Supplier Thermo Scientific Chemicals Supplier No. 02112018

CAS: 12014-56-1 | CeH8O4 | 212.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12014-56-1
Molecular Formula CeH8O4
Molecular Weight (g/mol) 212.18
MDL Number MFCD00010927
InChI Key DPUCLPLBKVSJIB-UHFFFAOYSA-N
SMILES O.O.O.O.[Ce]

Specifications

Color Brown to Yellow
Physical Form Lump/Powder
Quantity 50 g
Formula Weight 172.12 (anhydrous)
Odor Odorless
Chemical Name or Material Cerium(IV) oxide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation DOT Class: 8; Packaging Group: III
EINECSNumber 234-599-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only