missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) oxide, 96% (REO)

Catalog No. p-7051160
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4058130 250 g
AA40581A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4058130 Supplier Thermo Scientific Chemicals Supplier No. 04058130

CAS: 1306-38-3 | CeO2 | 172.114 g/mol

Cerium(IV) oxide is used in polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Polishing and decolorizing glass, in heat-resistant alloy coatings, or infrared filters to prevent reflection, and as a catalyst for organic reactions

Chemical Identifiers

CAS 1306-38-3
Molecular Formula CeO2
Molecular Weight (g/mol) 172.114
MDL Number MFCD00010933
InChI Key CETPSERCERDGAM-UHFFFAOYSA-N
Synonym ceric oxide, cerium dioxide, cerium iv oxide, ceria, cerium oxide, ceric dioxide, cerium 4+ oxide, needlal, nidoral, opaline
PubChem CID 73963
ChEBI CHEBI:79089
IUPAC Name dioxocerium
SMILES O=[Ce]=O

Specifications

Melting Point 2600°C
Color Yellow
Physical Form Powder
Quantity 250 g
Merck Index 14,1989
Solubility Information Insoluble in water; Soluble in H2SO4 and HNO3
Formula Weight 172.12
Percent Purity 96%
Odor Odorless
Density 7.132 g/mL
Assay (REO)
Chemical Name or Material Cerium(IV) oxide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 215-150-4
RTECSNumber FK6310000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only