missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min

CAS: 10378-47-9 | (NH4)4Ce(SO4)4·2H2O

Supplier:  Thermo Scientific Chemicals 040194A1

 View more versions of this product

Catalog No. AA40194A1


Only null left
Explore available promotions

Description

Description

Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium(IV) ammonium sulfate dihydrate
For analysis ACS
10378-47-9
1 kg
(NH4)4Ce(SO4)4·2H2O
Ammonium cerium sulfate
632.56 (596.53 Anhydrous)
Yellow crystalline
130°C
Yellow
Crystalline
94% min.
MFCD00149417
Soluble in water and insoluble in ethanol.
ACS

Chemical Identifiers

10378-47-9
(NH4)4Ce(SO4)4·2H2O
MFCD00149417
Ammonium cerium sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335-H500

EINECSNumber : 231-567-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only