missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium nitrate, ACS, 98.5% min

Catalog No. p-3257446
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3325422 100 g
AA3325436 500 g
AA33254A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3325422 Supplier Thermo Scientific Chemicals Supplier No. 03325422
Only null left

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins | CAS: 16774-21-3 | (NH4)2Ce(NO3)6

Analytical chemistry, as an oxidant for organic compounds, as a polymerization catalyst for olefins, and as a scavenger in the manufacture of azides. Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16774-21-3
Molecular Formula (NH4)2Ce(NO3)6
MDL Number MFCD00151121

Specifications

Physical Form Granules
Quantity 100 g
Assay Percent Range 98.5%
Solubility Information Soluble in water and alcohol.
Formula Weight 548.23
Grade ACS
Chemical Name or Material Cerium(IV) ammonium nitrate
Hazard Category H272-H290-H302-H314-H317-H335
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: II
EINECSNumber 240-827-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only