missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) ammonium nitrate, 98+%

Catalog No. AAA1288230
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1288230 250 g
AAA1288218 50 g
AAA128820B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1288230 Supplier Thermo Scientific Chemicals Supplier No. A1288230

Cerium(IV) ammonium nitrate, >-98%, H8CeN8O18, CAS Number-16774-21-3, Ammonium cerium(IV) nitrate; CAN, 1000g, Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S., 240-827-6, 548.23, H272-H290-H302-H314-H317-H335 | CAS: 16774-21-3 | H8CeN8O18

Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(IV) ammonium nitrate is widely utilized as an oxidant for many functional groups like alcohols, phenols and ethers as well as C-H bonds. It acts as a deprotection reagent for alcohol in organic synthesis. It is useful in the synthesis of heterocycles like quinoxaline derivatives which is used in dyes, organic semiconductors and DNA cleaving agents. It is an active component of chrome etchant, which is used in the production of photomasks and liquid crystal displays.

Solubility
Soluble in water and alcohol.

Notes
Store in cool place. Hygroscopic. Incompatible with strong reducing agents and powdered metals.

Chemical Identifiers

CAS 16774-21-3
Molecular Formula H8CeN8O18
MDL Number MFCD00151121
Synonym Ammonium cerium(IV) nitrate; CAN

Specifications

Quantity 250 g
Linear Formula (NH4)2Ce(NO3)6
UN Number UN1477
Merck Index 14,1992
Solubility Information Soluble in water and alcohol.
Formula Weight 548.23
Percent Purity ≥98%
Odor Pungent
Chemical Name or Material Cerium(IV) ammonium nitrate
Hazard Category H272-H290-H302-H314-H317-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber 240-827-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only