missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) Trifluoromethanesulfonate 90.0+%, TCI America™

Catalog No. p-7137022
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T19185G 5 g
T191825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T19185G Supplier TCI America Supplier No. T19185G
Only null left

Chemical Identifiers

CAS 76089-77-5
Molecular Formula C3CeF9O9S3
Molecular Weight (g/mol) 587.31
MDL Number MFCD02093899
InChI Key PHSMPGGNMIPKTH-UHFFFAOYSA-K
Synonym cerium iii trifluoromethanesulfonate, cerium iii triflate, trifluoromethanesulfonic acid cerium iii salt, cerium trifluoromethanesulfonate, cerous triflate, ce otf 3, 3cf3o3s.ce, acmc-209p1w, ksc491c9f
PubChem CID 2733941
IUPAC Name cerium(3+) tritrifluoromethanesulfonate
SMILES [Ce+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 587.31
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Cerium(III) Trifluoromethanesulfonate