missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) trifluoromethanesulfonate, 98%

Catalog No. AAL2025106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2025106 5 g
AAL2025114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2025106 Supplier Thermo Scientific Chemicals Supplier No. L2025106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 76089-77-5 | C3CeF9O9S3 | 587.31 g/mol

It is employed as intermediate for organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for organic synthesis and pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 76089-77-5
Molecular Formula C3CeF9O9S3
Molecular Weight (g/mol) 587.31
MDL Number MFCD02093899
InChI Key PHSMPGGNMIPKTH-UHFFFAOYSA-K
Synonym cerium iii trifluoromethanesulfonate, cerium iii triflate, trifluoromethanesulfonic acid cerium iii salt, cerium trifluoromethanesulfonate, cerous triflate, ce otf 3, 3cf3o3s.ce, acmc-209p1w, ksc491c9f
PubChem CID 2733941
IUPAC Name cerium(3+);trifluoromethanesulfonate
SMILES [Ce+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point >300°C
Density 1.7
Linear Formula (CF3SO3)3Ce
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 587.33
Percent Purity 98%
Chemical Name or Material Cerium(III) trifluoromethanesulfonate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only