Learn More
Cerium(III) sulfate octahydrate, REacton™, 99.999% (metals basis)
CAS: 10450-59-6 | Ce2(SO4)3·8H2O
$308.93 - $1248.07
Chemical Identifiers
| CAS | 10450-59-6 |
|---|---|
| Molecular Formula | Ce2(SO4)3·8H2O |
| MDL Number | MFCD00149641 |
Description
Cerium(III) sulfate octahydrate used in waterproofing, mildew-proofing, and in dyeing and printing textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10450-59-6 | |
| Ce2(SO4)3·8H2O | |
| MFCD00149641 |
Specifications
| 10450-59-6 | |
| 50 g | |
| MFCD00149641 | |
| Soluble in water. | |
| 99.999% | |
| 2.886 g/mL | |
| (metals basis) |
| Crystalline | |
| Ce2(SO4)3·8H2O | |
| 14,2002 | |
| 712.55 (568.42 Anhydrous) | |
| Reacton | |
| Cerium (III) sulfate octahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only