missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) phosphate, 99% min
Description
Intermediate product in cerium-based technique for staining lysosomal enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Physical Form | Powder |
| Quantity | 500 g |
| Odor | Odorless |
| Density | 5.22 g/mL |
| Linear Formula | CePO4 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 235.09 |
| Percent Purity | ≤99% |
| Chemical Name or Material | Cerium(III) phosphate |
Safety and Handling
| EINECSNumber | 236-637-8 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only