missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) perchlorate hexahydrate, Reagent Grade

Catalog No. p-7048848
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1165418 50 g
AA1165430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1165418 Supplier Thermo Scientific Chemicals Supplier No. 01165418
Only null left

CAS: 14017-47-1 | CeCl3O12 | 438.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14017-47-1
Molecular Formula CeCl3O12
Molecular Weight (g/mol) 438.454
MDL Number MFCD00150205
InChI Key LJBTWTBUIINKRU-UHFFFAOYSA-K
Synonym cerium 3+ perchlorate, cerium perchlorate, cerous perchlorate, cerium 3+ triperchlorate, perchloric acid, cerium 3+ salt, perchloric acid, cerium 3+ salt 3:1, acmc-1bs3z, cerium iii perchlorate solution, perchloric acid cerium iii salt, perchloric acid,cerium 3+ salt 3:1
PubChem CID 166978
IUPAC Name cerium(3+);triperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ce+3]

Specifications

Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Formula Weight 546.56 (438.47 Anhydrous)
Grade Reagent
Odor Odorless
Sensitivity Moisture sensitive
Chemical Name or Material Cerium(III) perchlorate hexahydrate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-836-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only