missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) oxalate hydrate, REacton™, 99.9% (REO)

Catalog No. p-7049752
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2112214 25 g
AA2112222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA2112214 Supplier Thermo Scientific Chemicals Supplier No. 02112214
Only null left

CAS: 15750-47-7 | C6Ce2O12 | 544.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15750-47-7
Molecular Formula C6Ce2O12
Molecular Weight (g/mol) 544.29
MDL Number MFCD00013082
InChI Key ZMZNLKYXLARXFY-UHFFFAOYSA-H
Synonym cerium iii oxalate hydrate, cerium iii oxalatehydrate, cerium iii oxalate decahydrate
PubChem CID 91998046
SMILES [Ce+3].[Ce+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 25 g
UN Number UN3288
Merck Index 14,2001
IUPAC Name dicerium(3+) trioxalate
Formula Weight 544.29 (Anhydrous)
Percent Purity 99.9%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Cerium(III) oxalate hydrate
Show More Show Less
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only