missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) nitrate hydrate, REacton™, 99.998% (metals basis)

Catalog No. AA4318122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4318122 100 g
AA4318114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4318122 Supplier Thermo Scientific Chemicals Supplier No. 04318122

Ce(NO3)3·xH2O (x ≈ 6-7)

Cerium(III) nitrate hydrate is used as an analytical reagent. It acts as a catalyst in petrochemical industry and in the pharmaceutical industry. Further, it is a chemical that helps burn tissue recover in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

Molecular Formula Ce(NO3)3·xH2O (x ≈ 6-7)
MDL Number MFCD00010926

Specifications

Color White
Physical Form Crystalline
Quantity 100 g
Assay Percent Range 99.998%
Solubility Information Soluble in water,alcohol and acetone
Formula Weight 326.13 (Anhydrous)
Odor Acidic
Sensitivity Hygroscopic
Appearance White crystalline
Chemical Name or Material Cerium(III) nitrate hydrate, 150°C - 3H2O
Hazard Category H272-H315-H318-H350
Precautionary Statement P201-P202-P220-P221-P264b-P271-P280-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P370+P380+P375-P372-P501c
DOTInformation DOT Class: 5.1, Packaging Group: III
EINECSNumber 233-297-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only