missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride hydrate, REacton™, 99.9% (REO)

CAS: 19423-76-8 | CeCl3 | 246.47 g/mol

$109.10 - $317.75

Chemical Identifiers

CAS 19423-76-8
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149633
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym Cerous chloride
IUPAC Name cerium(3+) hydrate trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1132518
View Documents
Thermo Scientific Chemicals
01132518
50 g
Each for $109.10
Only null left
 
AA1132530
View Documents
Thermo Scientific Chemicals
01132530
250 g
Each for $317.75
Only null left
 
Description

Description

Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

19423-76-8
246.47
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium(3+) hydrate trichloride
CeCl3
MFCD00149633
Cerous chloride
[Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

White
Crystalline Aggregates
99.9%
MFCD00149633
Soluble in water and strong mineral acids.
[Cl-].[Cl-].[Cl-].[Ce+3]
246.47
Air sensitive; Hygroscopic
19423-76-8
50 g
CeCl3
Cerous chloride
VYLVYHXQOHJDJL-UHFFFAOYSA-K
cerium(3+) hydrate trichloride
246.48 (Anhydrous)
Cerium(III) chloride hydrate, Rare Earth Oxides Content
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 232-227-8

RTECSNumber : FK5150000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only