missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride hydrate, REacton™, 99.9% (REO)

Catalog No. AA1132518
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1132518 50 g
AA1132530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1132518 Supplier Thermo Scientific Chemicals Supplier No. 01132518

CAS: 19423-76-8 | CeCl3 | 246.47 g/mol

Cerium(III) chloride hydrate is used as an important raw material the preparation of other cerium salts such as cerium(III) trifluoromethanesulfonate. It is used in the conversion of esters to allylsilanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19423-76-8
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149633
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym Cerous chloride
IUPAC Name cerium(3+) hydrate trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 50 g
Assay Percent Range 99.9%
Solubility Information Soluble in water and strong mineral acids.
Formula Weight 246.48 (Anhydrous)
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material Cerium(III) chloride hydrate, Rare Earth Oxides Content
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-227-8
RTECSNumber FK5150000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only