Learn More
Cerium(III) chloride heptahydrate, 99%
CAS: 18618-55-8 | CeCl3·7H2O
Supplier: Thermo Scientific Chemicals A129470B
Description
Cerium(III) chloride heptahydrate is used in the preparation of allylsilanes from esters. It is used as a reducing agent in organic synthesis in place of sodium borohydride. In Luche reaction, carvone gives selectively allylic alcohol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCerium(III) chloride heptahydrate is used in the preparation of allylsilanes from esters. It is used as a reducing agent in organic synthesis in place of sodium borohydride. In Luche reaction, carvone gives selectively allylic alcohol.
Solubility
Soluble in alcohol and acetone. Slightly soluble in tetrahydrofuran. Insoluble in cold water.
Notes
Air sensitive. Hygroscopic. Incompatible with strong acids and strong oxidizing agents.
Specifications
| Cerium(III) chloride heptahydrate | |
| 1000 g | |
| MFCD00149634 | |
| Cerous chloride heptahydrate | |
| 372.60 (246.48 Anhydrous) | |
| Odorless |
| 18618-55-8 | |
| CeCl3·7H2O | |
| 14,1997 | |
| Soluble in alcohol,water,and acetone. Slightly soluble in tetrahydrofuran. | |
| 99% | |
| Air sensitive; Hygroscopic |
Chemical Identifiers
| 18618-55-8 | |
| CeCl3·7H2O | |
| MFCD00149634 | |
| Cerous chloride heptahydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 606-073-0
RTECSNumber : FK5100000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only