missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) carbonate hydrate, REacton™, 99.9% (REO)

Catalog No. p-4824657
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1529522 100 g
AA1529536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1529522 Supplier Thermo Scientific Chemicals Supplier No. 01529522

CAS: 54451-25-1 | C3Ce2O9 | 460.26 g/mol

Raw material of various high purity cerium complexes and high grad polishing powder. Cerium Carbonate is used to decolorize glass by keeping iron in its ferrous state.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54451-25-1
Molecular Formula C3Ce2O9
Molecular Weight (g/mol) 460.26
MDL Number MFCD00149635
InChI Key GHLITDDQOMIBFS-UHFFFAOYSA-H
Synonym cerium iii carbonate hydrate, acmc-20alhm, cerous carbonate hydrate, ksc267s0r, ce2 co3 3.xh2o, cerium iii carbonate hydrate, reacton, cerium 3+ carbonate-water 2/3/1, dicerium 3+ hydrate tricarbonate, cerium iii carbonate hydrate, reacton reo
PubChem CID 16212049
SMILES [Ce+3].[Ce+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

Specifications

Melting Point 848°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.9%
Solubility Information Insoluble in water. Soluble indilute mineral acids.
IUPAC Name dicerium(3+) tricarbonate
Formula Weight 460.27 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Cerium(III) carbonate hydrate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-655-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only