missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerium(III) carbonate hydrate, REacton™, 99.9% (REO)
Description
Raw material of various high purity cerium complexes and high grad polishing powder. Cerium Carbonate is used to decolorize glass by keeping iron in its ferrous state.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 848°C |
| Color | White |
| Physical Form | Crystals |
| Quantity | 500 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Assay Percent Range | 99.9% |
| Solubility Information | Insoluble in water. Soluble indilute mineral acids. |
| IUPAC Name | cerium(3+);tricarbonate;hydrate |
| Formula Weight | 460.27 (anhydrous) |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 208-655-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only