missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) Acetate Hydrate 98.0+%, TCI America™

Catalog No. C297425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C297425G 25 g
C2974500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C297425G Supplier TCI America Supplier No. C297425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 206996-60-3
Molecular Formula C6H9CeO6
Molecular Weight (g/mol) 317.25
MDL Number MFCD00051060
InChI Key VGBWDOLBWVJTRZ-UHFFFAOYSA-K
Synonym Acetic Acid Cerium(III) Salt
IUPAC Name cerium(3+) triacetate
SMILES [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Melting Point 308°C
Color White-Yellow
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 317.25
Percent Purity ≥98.0% (T)
Chemical Name or Material Cerium(III) Acetate Hydrate
EINECSNumber (2)-0705
RTECSNumber FK4900000
TSCA Yes