Learn More
Cerium(III) acetate hydrate, 99.995% (metals basis)
CAS: 206996-60-3 | C6H9CeO6 | 317.25 g/mol
$133.64 - $1957.73
Chemical Identifiers
| CAS | 206996-60-3 |
|---|---|
| Molecular Formula | C6H9CeO6 |
| Molecular Weight (g/mol) | 317.25 |
| MDL Number | MFCD00051060 |
| InChI Key | VGBWDOLBWVJTRZ-UHFFFAOYSA-K |
| IUPAC Name | cerium(3+) triacetate |
| SMILES | [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA4443309
|
Thermo Scientific Chemicals
04443309 |
10 g |
Each for $133.64
|
|
|||||
|
AA4443318
|
Thermo Scientific Chemicals
04443318 |
50 g |
Each for $474.86
|
|
|||||
|
AA4443330
|
Thermo Scientific Chemicals
04443330 |
250 g |
Each for $1,957.73
|
|
|||||
Description
Cerium(III) acetate hydrate is used in thin fim buffer layers suitable for high κ dielectrics by sol-gel route. And it is also used as a catalyst for auto-oxidation of cresols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 206996-60-3 | |
| 317.25 | |
| VGBWDOLBWVJTRZ-UHFFFAOYSA-K | |
| [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O |
| C6H9CeO6 | |
| MFCD00051060 | |
| cerium(3+) triacetate |
Specifications
| White | |
| Crystalline | |
| C6H9CeO6 | |
| MFCD00051060 | |
| VGBWDOLBWVJTRZ-UHFFFAOYSA-K | |
| cerium(3+) triacetate | |
| 317.26 (Anhydrous) | |
| Cerium (III) acetate hydrate |
| 206996-60-3 | |
| 10 g | |
| Ce(C2H3O2)3·xH2O | |
| Soluble in water( 3.5g/L). | |
| [Ce+3].CC([O-])=O.CC([O-])=O.CC([O-])=O | |
| 317.25 | |
| 99.995% | |
| (metals basis) |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only