missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) 2-ethylhexanoate, 49% in 2-ethylhexanoic acid, Ce 12%

CAS: 56797-01-4 | C24H45CeO6 | 569.73 g/mol

Supplier:  Thermo Scientific Chemicals 04045136

 View more versions of this product

Catalog No. AA4045136


Only null left

Description

Description

Used as catalyst, paint dryer, plastic Additive. As the the precursor in preparation of inorganic nanoparticles in oil-in-water microemulsions. The Toluene-based precursor solution was composed by mixing gadolinium (III) 2-ethylhexanoate and cerium (III) 2-ethylhexanoate to achieve the right stoichiometry. For the ceria/zirconia precursor, cerium(III) 2-ethylhexanoate and zirconium(IV) 2-ethylhexanoate were mixed to result in a metal molar ratio Ce/Zr of 1:1 and diluted with xylene to a total metal. Thin films of CeO 2 were prepared by a photochemical method using thin amorphous films of cerium(III) 2-ethylhexanoate as the precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cerium(III) 2-ethylhexanoate
1.08 g/mL
C24H45CeO6
500 g
Insoluble in water.
[Ce+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
569.73
569.74
Viscous Liquid
56797-01-4
>110°C (230°F)
MFCD00014004
cerium iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt, cerium tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt 1:?, hexanoic acid, 2-ethyl-, cerium 3+ salt 3:1, hexanoic acid, 2-ethyl-, cerium 3+ salt, cerium 2-ethylhexanoate, cerous 2-ethylhexanoate, 2-ethylhexanoic acid, cerium salt, cerium hex-cem
GGVUYAXGAOIFIC-UHFFFAOYNA-K
cerium(3+);2-ethylhexanoate
62122
49% in 2-ethylhexanoic acid, Ce 12%

Chemical Identifiers

56797-01-4
569.73
GGVUYAXGAOIFIC-UHFFFAOYNA-K
62122
[Ce+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
C24H45CeO6
MFCD00014004
cerium iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt, cerium tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt 1:?, hexanoic acid, 2-ethyl-, cerium 3+ salt 3:1, hexanoic acid, 2-ethyl-, cerium 3+ salt, cerium 2-ethylhexanoate, cerous 2-ethylhexanoate, 2-ethylhexanoic acid, cerium salt, cerium hex-cem
cerium(3+);2-ethylhexanoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361
Suspected of damaging fertility or the unborn child.

P201-P202-P281-P308+P313-P501c

H361d

EINECSNumber : 260-386-3

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only