missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) 2-ethylhexanoate, 49% in 2-ethylhexanoic acid, Ce 12%

Catalog No. AA4045122
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4045122 100 g
AA4045136 500 g
AA40451A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4045122 Supplier Thermo Scientific Chemicals Supplier No. 04045122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56797-01-4 | C24H45CeO6 | 569.73 g/mol

Used as catalyst, paint dryer, plastic Additive. As the the precursor in preparation of inorganic nanoparticles in oil-in-water microemulsions. The Toluene-based precursor solution was composed by mixing gadolinium (III) 2-ethylhexanoate and cerium (III) 2-ethylhexanoate to achieve the right stoichiometry. For the ceria/zirconia precursor, cerium(III) 2-ethylhexanoate and zirconium(IV) 2-ethylhexanoate were mixed to result in a metal molar ratio Ce/Zr of 1:1 and diluted with xylene to a total metal. Thin films of CeO 2 were prepared by a photochemical method using thin amorphous films of cerium(III) 2-ethylhexanoate as the precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56797-01-4
Molecular Formula C24H45CeO6
Molecular Weight (g/mol) 569.73
MDL Number MFCD00014004
InChI Key GGVUYAXGAOIFIC-UHFFFAOYNA-K
Synonym cerium iii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt, cerium tris 2-ethylhexanoate, hexanoic acid, 2-ethyl-, cerium salt 1:?, hexanoic acid, 2-ethyl-, cerium 3+ salt 3:1, hexanoic acid, 2-ethyl-, cerium 3+ salt, cerium 2-ethylhexanoate, cerous 2-ethylhexanoate, 2-ethylhexanoic acid, cerium salt, cerium hex-cem
PubChem CID 62122
IUPAC Name cerium(3+);2-ethylhexanoate
SMILES [Ce+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O

Specifications

Density 1.08 g/mL
Flash Point >110°C (230°F)
Quantity 100 g
Solubility Information Insoluble in water.
Formula Weight 569.74
Concentration or Composition (by Analyte or Components) 49% in 2-ethylhexanoic acid, Ce 12%
Physical Form Viscous Liquid
Chemical Name or Material Cerium(III) 2-ethylhexanoate
Hazard Category H361d
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 260-386-3
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only