missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Pentylone Hydrochloride, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001860
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001860 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001860 Supplier Merck Emd Millipore Supplier No. P0861ML
Only null left

Certified reference material

Developed in the 1960's, pentylone is a newer bath salt cathinone and stimulant sold in a blend called 'NRG-1' with other bath salts such as flephedrone, MDPBP, MDPV, and MPPP. Pentylone has also been found in a separate blend with MPPP called 'NRG-3.'

Specifications

CAS 67-56-1, 17763-01-8
Linear Formula C13H17NO3 · HCl
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol (as free base)
Synonym Pentylone hydrochloride solution
Formula Weight 271.74
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation