missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Mitragynine-D3 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001660
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001660 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001660 Supplier Merck Emd Millipore Supplier No. M1821ML
Only null left

Certified reference material

An internal standard suitable for quantitation of mitragynine in biological samples using mass spectrometry-based methods. Mitragynine, commonly known as Kratom, is the most abundant alkaloid in the plant Mitragynine speciosa. Sold over the internet as herbal plant extracts, Kratom is noted for its psychoactive, stimulant, and sedative effects depending on the dosage level. According to DEA's Drugs and Chemicals of Concern page, mitragynine use can lead to addiction with adverse effects ranging from anorexia and weight loss to hallucinations and delusion.

Specifications

CAS 67-56-1
Linear Formula C23H27D3N2O4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 401.51
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation