missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Methedrone Hydrochloride Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001636
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001636 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001636 Supplier Merck Emd Millipore Supplier No. M1471ML
Only null left

Certified reference material

Methedrone is a synthetic stimulant of the phenethylamine, amphetamine, and cathinone classes. Significant abuse of this new designer drug, also known as 4-methoxymeth-cathinone, methoxyphedrine, or bk-PMMA, has been reported within the last year in Sweden, the UK, Poland and states including Louisiana and Kentucky.

Specifications

Packaging Ampule
CAS 879665-92-6
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C11H16ClNO2
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Synonym 4-Methoxymethcathinone hydrochloride
Quantity 1 mL
Formula Weight 229.7

CERILLIANT is a registered trademark of Cerilliant Corporation