missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Ketamine-D4 Hydrochloride Solution, 100 μg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001522
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001522 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001522 Supplier Merck Emd Millipore Supplier No. K0031ML
Only null left

Certified reference material

This stable-labeled internal standard is suitable for quantitation of ketamine levels in GC/MS or LC/MS applications from clinical toxicology and forensic analysis to urine drug testing and isotope dilution methods. Ketamine, marketed under trade names Ketanest, Ketaset and Ketalar, is an anesthetic used in human and veterinary medicine. The drug is also abused at clubs, raves, or parties for its noted hallucinogenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression.

Specifications

CAS 67-56-1, 1246815-97-3
Linear Formula C13D4H12ClNO · HCl
MDL Number MFCD28038722
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol (as free base)
Formula Weight 278.21
Packaging Ampule
Quantity 1 mL

CERILLIANT is a registered trademark of Cerilliant Corporation