missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Heroin-D9 Solution, 100 μg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001422
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001422 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001422 Supplier Merck Emd Millipore Supplier No. H0361ML
Only null left

Certified reference material

A stable-labeled internal standard with suitable uses in LC/MS and GC/MS application for quantitation of heroin levels in clinical toxicology, forensic analysis and urine drug testing applications. Heroin, also known as diacetylmorphine, is an opiate analgesic most commonly used as an illcit recreational drug.

Specifications

CAS 75-05-8, 1338713-49-7
Linear Formula C21H14NO5D9
MDL Number MFCD00798480
UN Number UN 1648 3 / PGII
Concentration 100 μg/mL in acetonitrile
Formula Weight 378.47
Packaging Ampule
Quantity 1 mL
Flash Point 2°C (35.6°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation