missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Heroin-D9 Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
A stable-labeled internal standard with suitable uses in LC/MS and GC/MS application for quantitation of heroin levels in clinical toxicology, forensic analysis and urine drug testing applications. Heroin, also known as diacetylmorphine, is an opiate analgesic most commonly used as an illcit recreational drug.
Specifications
Specifications
| CAS | 75-05-8, 1338713-49-7 |
| Linear Formula | C21H14NO5D9 |
| MDL Number | MFCD00798480 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 378.47 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation