missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Ginger Gingerols and Shogaols Mix Solution, certified reference material, Sold by MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 75-05-8, 23513-15-7 |
| Linear Formula | C17H26O4 |
| Formula Weight | 294.39 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 6°C (42.8°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation