missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Flubromazolam Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001371
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001371 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001371 Supplier Merck Emd Millipore Supplier No. F0471ML
Only null left

Certified reference material

Flubromazolam is a highly potent benzodiazepine drug also sold as designer drug due to its ability to produce strong sedation and amnesia. Flubromazolam reportedly has a high addiction potential and risk of overdose. This Certified Spiking Solution™ is suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing.

Specifications

CAS 67-56-1, 612526-40-6
Linear Formula C17H12BrFN4
MDL Number MFCD29036758
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 1.0 mg/mL in methanol
Formula Weight 371.21
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation