missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Estazolam-d5 Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001332
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001332 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001332 Supplier Merck Emd Millipore Supplier No. E9031ML
Only null left

Certified reference material

A stable-labeled internal standard suitable for quantitation of estazolam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Estazolam is a benzodiazepine prescribed for the short-term treatment of insomnia. Sold under trade names such as ProSom or Eurodin, estazolam is often abused recreationally to achieve a high or when long-term use is continued against physician advice.

Specifications

CAS 67-56-1, 170082-16-3
Linear Formula C16D5H6ClN4
MDL Number MFCD00798476
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Concentration 100 μg/mL in methanol
Formula Weight 299.77
Packaging Ampule
Quantity 1 mL
Flash Point 9.7°C (49.5°F) - closed cup

CERILLIANT is a registered trademark of Cerilliant Corporation