missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Epitestosterone Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Description
Epitestosterone is a stereoisomer of the hormone testosterone. Epitestosterone is often used to illicitly mask high testosterone levels in athletic drug tests. This Certified Snap-N-Spike™ Solution is applicable for use in LC/MS or GC/MS applications for sports testing, urine drug testing or forensic analysis.
Specifications
Specifications
| CAS | 75-05-8, 481-30-1 |
| Linear Formula | C19H28O2 |
| MDL Number | MFCD00067129 |
| UN Number | UN 1648 3 / PGII |
| Concentration | 1.0 mg/mL in acetonitrile |
| Formula Weight | 288.42 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 2°C (35.6°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation