missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CERILLIANT™ Diethylpropion-D10 Hydrochloride Solution, 100 μg/mL in Methanol (with 1% 1 M HCl, Sold by MilliporeSigma™ Supelco™
Description
Also known as diethylcathinone or amfepramone, diethylpropion is a stimulant and appetite suppressant drug of the phenethylamine, amphetamine, and cathinone chemical classes. This new internal standard is suitable for numerous GC/MS and LC/MS applications from forensic analysis and clinical toxicology testing to urine drug testing and isotope dilution methods
Specifications
Specifications
| CAS | 67-56-1, 1189500-62-6 |
| Linear Formula | C13H9D10NO · HCl |
| MDL Number | MFCD11977883 |
| UN Number | UN 1230 - class 3 - PG 2 - Methanol, solution |
| Concentration | 100 μg/mL in methanol (with 1% 1 M HCl |
| Formula Weight | 251.82 |
| Packaging | Ampule |
| Quantity | 1 mL |
| Flash Point | 9.7°C (49.5°F) - closed cup |
CERILLIANT is a registered trademark of Cerilliant Corporation