missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Carfentanil Oxalate Solution, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Catalog No. 01001115
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001115 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001115 Supplier Merck Emd Millipore Supplier No. C1621EA
Only null left

Certified reference material

A certified Snap-N-Spike™ solution suitable for use as starting material in calibrators and controls for fentanyl LC/MS or GC/MS testing methods in clinical toxicology, urine drug testing, prescription monitoring, or forensic analysis applications. Marketed under the trade name Wildnil as a general anesthetic for large animals, carfentanil hit the illicit market in 2016 as a new NPS drug or in combination with heroin. Carfentanil, also known as 4-carbomethoxyfentanyl, is a fentanyl analog 10,000 times more potent than morphine, making it one of the most potent synthetic opioids available.

Specifications

Packaging Ampule
CAS 67-56-1
Concentration 100 μg/mL in methanol
Linear Formula C24H30N2O3 · C2H2O4
MDL Number MFCD01317854
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 0.5 mL
Formula Weight 484.54

CERILLIANT is a registered trademark of Cerilliant Corporation