missing translation for 'onlineSavingsMsg'
Learn More

CERILLIANT™ Acetyl norfentanyl oxalate Solution, 1.0 mg/mL in Methanol (as free base), Sold by MilliporeSigma™ Supelco™

Catalog No. 01001008
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01001008 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01001008 Supplier Merck Emd Millipore Supplier No. A1151ML
Only null left

Certified reference material

Acetyl norfentanyl is a major metabolite of acetyl fentanyl, a designer drug and fentanyl analog with a potency 40 times greater than heroin and 80 times greater than morphine. In June, 2013, the Centers for Disease Control (CDC) issued a health alert for acetyl fentanyl, reporting 14 overdose deaths in Rhode Island. This certified solution standard is suitable for a variety of LC/MS or GC/MS testing applications from forensic analysis, clinical toxicology, and pain prescription monitoring to urine drug testing and pharmaceutical research.

Specifications

Packaging Ampule
CAS 67-56-1
Flash Point 9.7°C (49.5°F) - closed cup
Concentration 1.0 mg/mL in methanol (as free base)
Linear Formula C13H18N2O · C2H2O4
UN Number UN 1230 - class 3 - PG 2 - Methanol, solution
Quantity 1 mL
Formula Weight 308.33

CERILLIANT is a registered trademark of Cerilliant Corporation