missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate), Reagents

Catalog No. 01338056
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01338056 100 g
01338055 2 kg
01338054 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338056 Supplier Labchem Inc Supplier No. C1171520100C2
Only null left

Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 16774-21-3
Molecular Formula CeH8N8O18
Molecular Weight (g/mol) 548.22
InChI Key WIBGOERAEYJBOT-UHFFFAOYSA-N
Synonym Ammonium Cerium(IV) Nitrate, Ammonium Hexanitratocerate (IV)
IUPAC Name λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
SMILES N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Orange/Red or Orange/Yellow
pH 1
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.49
Chemical Name or Material Ceric Ammonium Nitrate, ACS Reagent (Ammonium Cerium (IV) Nitrate)