missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Molybdate Solution (contains H2SO4) [for TLC Stain], TCI America™

$182.68 - $508.89

Chemical Identifiers

MDL Number MFCD06797081
Synonym CAM Solution
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C1794100ML
View Documents
TCI America
C1794100ML
100 mL
Each for $182.68
Only null left
 
C1794500ML
View Documents
TCI America
C1794500ML
500 mL
Each for $508.89
Only null left
 
Description

Description

Specifications

Chemical Identifiers

MFCD06797081
CAM Solution

Specifications

Yellow
100 mL
CAM Solution
Ceric Ammonium Molybdate Solution (contains H2SO4) [for TLC Stain]
MFCD06797081
3264
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'