missing translation for 'onlineSavingsMsg'
Learn More

Ceric Ammonium Molybdate Solution (contains H2SO4) [for TLC Stain], TCI America™

Catalog No. C1794100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1794100ML 100 mL
C1794500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1794100ML Supplier TCI America Supplier No. C1794100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD06797081
Synonym CAM Solution

Specifications

Color Yellow
Quantity 100 mL
UN Number 3264
Physical Form Liquid
Chemical Name or Material Ceric Ammonium Molybdate Solution (contains H2SO4) [for TLC Stain]