missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Ceramic Magnets

Catalog No. S15906
Change view
Click to view available options
Dimensions (L x W x H):
50 x 10 x 7 mm
70 x 15 x 8 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S15906 50 x 10 x 7 mm
S15888 70 x 15 x 8 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15906 Supplier United Scientific™ Supplier No. CBM050

Simple strong magnets for the classroom.

  • We offer a variety of economical ceramic bar, disc, and ring shaped magnets.
  • Magnets are unpainted, except where noted.

Specifications

Material Ceramic
For Use With (Application) Magnetic Demonstrations
Grade 5+
Product Type Magnet
Dimensions (L x W x H) 50 x 10 x 7 mm