missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ CentriVap™ Concentrator Accessory, High Capacity Micro Rotor

Catalog No. 16108551
Change view
Click to view available options
Material:
Aluminum
Anodized Aluminum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
16108551 Aluminum
16108550 Anodized Aluminum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16108551 Supplier Labconco™ Supplier No. 7553501

For use with Labconco™ CentriVap™ Concentrators. Labconco™ CentriVap™ Concentrator Accessory, rotors are constructed of anodized aluminum and hold tubes at an angle to maximize surface area.

  • Available in standard, or with fluoropolymer resin coating that ensures resistance to acids
  • High capacity micro rotor holds up to 148 micro centrifuge tubes, 1.5mL or 2.0mL, and micro auto sample vials approx. 12mm in dia. x 35mm
  • High capacity micro rotors don't require lip on the tube and are perfect for vials

Warranty

One year

Specifications

Description 30.4 x 30.4 x 30.4cm (12 x 12 x 12 in.)
Material Aluminum
Type Micro Rotor
For Use With (Equipment) Labconco™ CentriVap™ Concentrator
Holds 1.5 mL or 2.0 mL Microcentrifuge Tubes, approx. 12 mm in dia. x 35 mm Micro Autosample Vials
Material (Coating) PTFE
Weight (English) 2.5 lb.
Weight (Metric) 1.1 kg