Learn More
Labconco CentriVap™ Cold Trap with Stainless Steel Lid
Designed to collect evaporated liquid from a CentriVap™ Concentrator, protecting the vacuum pump and lab environment from vapors and fumes.
$11306.04 - $11306.04
Specifications
Material Stainless Steel
Type Cold Trap
Plug Type North American
View More Specs
Products 2
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
12-199-541
View Documents
Labconco
8601110
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
12-199-509
View Documents
Labconco
8601100*
N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Brushed, stainless steel and powder-coated steel exterior
  • Illuminated temperature graph with endpoint indication
  • HCFC-free refrigerant system
  • 4 L stainless steel collector
  • Stainless steel lid with 2 quick-disconnect fittings for connection to a CentriVap™ concentrator
  • Stainless steel drain port (with corrosion-resistant, nylon valve)
  • -50 °C models feature a 1/3 hp compressor (-50 °C in less than 30 min.)
Specifications
Specifications
Stainless Steel
Cold Trap
North American
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'